missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Hyperscreen™ Intensifying Screen
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
Brand: Cytiva RPN1669
Code : GE
Additional Details : Weight : 1.00000kg
Description
- Light emission from calcium tungstate phosphor is fully compatible with Hyperfilm™ MP
- Use with Sensitize™ Pre-Flash Unit for even higher sensitivity and accurate quantitation
- Fine grain screens give clear band resolution
- Available in a full range of sizes
Specifications
8 x 10 in. | |
Hyperscreen intensifying screens are economical high-performance calcium tungstate screens. They can be used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results. | |
203 mm | |
254 mm |
Hyperscreen | |
7.99 in. | |
10 in. | |
Used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results |