missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ -XL Membranes

Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.

Brand:  Cytiva RPN203S

Code : GE

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10024394

  • £294.00 / 1 roll


Estimated Shipment
11-03-2021
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600μg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available

Specifications

Specifications

1 Roll
Nylon, Highly Positively Charged
3 m
Up to 600μg/cm2
Roll
20 cm
Radioactive probes
Videos
SDS
Documents