Learn More
Cytiva Amersham™ Hybond™ -XL Membranes
Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
£308.04
Specifications
For Use With (Equipment) | Radioactive probes |
---|---|
Binding Capacity | Up to 600μg/cm2 |
Material | Nylon, Highly Positively Charged |
Product Code | Brand | Length (Metric) | Width (Metric) | Format | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Format | Quantity | Price | Quantity & Availability | ||
12120960
|
Cytiva
RPN2020S |
20 cm | 20 cm | Sheet | 10 Sheets |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Greater concentration and more even distribution of positive charge than other charged nylon membranes
- Binding capacity for nucleic acids up to 600μg/cm2
- Efficient binding of small fragments
- Effective probe removal
- Suitable for reprobing
- Discs are also available
Specifications
Radioactive probes | |
Up to 600μg/cm2 | |
Nylon, Highly Positively Charged |