missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Amersham™ Hybond™ -XL Membranes

Use for optimal signal-to-noise ratios when using radioactively labeled probes. GE Healthcare Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.

Brand:  GE Healthcare RPN303S

Code : GE

Additional Details : Weight : 0.31100kg

 View more versions of this product

Product Code. 10712817

  • £355.00 / Each


Estimated Shipment
06-06-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600μg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available

Specifications

Specifications

1 Roll
3m
Nylon, Highly Positively Charged
Roll
30cm
Radioactive probes
Up to 600μg/cm2
Videos
Documentation

Technical Documents