missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Amersham™ Hybond™ PVDF Membranes: Sheets

Ideal for Western blotting applications where stripping and reprobing are necessary

Brand:  GE Healthcare 10600090

Code : GH

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 15209894

  • £140.00 / 25 sheets


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Individual sheets.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    Specifications

    Specifications

    25/Pk.
    PVDF
    10 × 10cm
    Ideal for Western blotting applications where stripping and reprobing are necessary
    Sheets (LFP)
    Videos