missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Amersham™ Hybond™ -NX Membranes

Neutral nylon membrane developed specifically for use with low hybridization buffer volumes and high-throughput applications

Brand:  GE Healthcare RPN303T

Code : GE

Additional Details : Weight : 1.42430kg

 View more versions of this product

Product Code. 10615545

  • £271.00 / Each


Estimated Shipment
19-02-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Specifications

Specifications

30cm
Radioactive probes
Up to 600μg/cm2
3m
Nylon, Highly Positively Charged
Roll
Videos
Documentation

Technical Documents