missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Amersham™ CyDye™ Antibody Labeling Kits

Convenient way to tag antibodies with CyDye fluorescent dyes

Brand:  GE Healthcare PA35001

Code : GE

Additional Details : CAS Number : 497-19-8 Weight : 1.42430kg

 View more versions of this product

Product Code. 10564195

  • £130.56 / 1 set


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Specifications

Specifications

For the conjugation of Proteins with Cy5 mono-functional dye
Contain all reagents required for 2 x 0.1mg reactions
Cy5 MAb Labeling Kit
Each
Cy5 mono-functional dye
7 to 7.3
2° to 8°C
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .