missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Amplify™ Fluorographic Reagent

Fast and convenient fluorographic reagent for 35S-, 14C- and 3H-labeled samples in gels

Brand:  Cytiva NAMP100

Code : GE

Additional Details : CAS Number : 55965-84-9 Weight : 1.00000kg


Product Code. 10094154

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Increases sensitivity of detection and significantly reduce exposure times required for weak beta emitters.

  • Increased detection efficiency 1000-fold for 3H and 15-fold for 35S and 14C
  • Less than two hours total preparation time
  • Can be used with acrylamide or agarose gels with no change in protocols
  • Odorless, inorganic and fully compatible with Sensitize™ Preflash unit and Hyperfilm™ MP

Specifications

Specifications

Fast and convenient fluorographic reagent for 35S-, 14C- and 3H-labeled samples in Gels
3.5 to 4.5
1000 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 1,2,4-Trimethyl-benzene5-sulphonic acid
  • UNKNOWN AQUATIC-Contains 5.9% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers