missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare 2-D Protein Extraction Buffers

Designed to produce high spot resolution for 2-D gel analysis

Brand:  GE Healthcare 28-9435-28

UNSPSC : 12161700

Code : GE

Additional Details : Weight : 1.00000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

 View more versions of this product

Product Code. 10626355

  • £52.99 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Specifications Description & Specifications

Specifications

White
Solid
171° to 175°C
50mL
Buffer VI with Diluent III
1X
Contains <8M Urea, <5M Thiourea, <5% CHAPS, <4% NDSB 201
76.11616
15° to 30°C

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P281-Use personal protective equipment as required.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .