missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare 2-D Protein Extraction Buffers

Designed to produce high spot resolution for 2-D gel analysis

Brand:  GE Healthcare 28-9435-25

Code : GE

Additional Details : Weight : 0.05000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

 View more versions of this product

Product Code. 10064884

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Convenient way to prepare quality protein lysates
  • Ready to mix buffers simplifies protein extraction preparation of cells and tissues
  • Efficient and reproducible total protein extraction with yield
  • Can also be used prior to 1-D electrophoresis, or for rehydrating IPG strips to 2-D electrophoresis
  • Each package contains sufficient amounts of reagents to prepare 50mL buffer

Specifications

Specifications

Clear
Liquid
50mL
Buffer III with Diluent III
1X
Contains <8M Urea, <5M Thiourea, <5% CHAPS, <5% ASB-16
15° to 30°C
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .