missing translation for 'onlineSavingsMsg'
Learn More

Mouse GDF-3 Propeptide Antibody, R&D Systems™

Rat Monoclonal Antibody

POA

Specifications

Antigen GDF-3
Clone 356613
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Description

Description

GDF-3 Monoclonal specifically detects GDF-3 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

GDF-3
LYOPH
Western Blot
Unconjugated
Rat
Mouse
Q3ZB19
9573
Mouse myeloma cell line NS0-derived recombinant mouse GDF-3 Propeptide Ser23-Lys250 Accession # Q3ZB19
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse GDF-3 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) GDF-1, rmGDF-1pro, rmGDF-3, rmGDF-5, rmGDF-6, rmGDF-7, rmGDF-8, or rmGDF-9 is observed.
356613
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2
GDF3
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only