missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-1 Propeptide Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | GDF-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
GDF-1 Polyclonal specifically detects GDF-1 in Mouse samples. It is validated for Western Blot.Specifications
GDF-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
DORV, DTGA3, embryonic growth/differentiation factor 1, GDF1, GDF-1, growth differentiation factor 1 | |
GDF1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P20863 | |
2657 | |
Mouse myeloma cell line NS0-derived recombinant mouse GDF-1 Propeptide Pro25-Leu235 Accession # P20863 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse GDF-1 Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant mouse (rm) GDF-1 and rmGDF-3 Propeptide is observed. |
For Research Use Only