missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ GC Column Phe-Sil Retention Gap

Brand:  Macherey Nagel™ 723711.25

UNSPSC : 41115700

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10738622

  • £560.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

GC Column Retention Gap
320°C
GC
1Each
0.8mm
25m
0.53mm
For all Solvents Except Methanol and Water