missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ GC Column CW Retention Gap

Brand:  Macherey-Nagel™ 723714.10

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10586861

  • £252.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.53 mm
0.8 mm
250°C
1 Each
GC Column Retention Gap
Solvents and Especially for Methanol and Water Analysis
10 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers