missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ GC Column CW Retention Gap

Brand:  Macherey-Nagel™ 723712.25

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10546691

  • £436.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

GC Column Retention Gap
250°C
GC
1Each
0.4mm
25m
0.25mm
For all Solvents and Especially for Methanol and Water
Videos