missing translation for 'onlineSavingsMsg'
Learn More

GBR 13069 dihydrochloride, Tocris Bioscience™

Potent dopamine uptake inhibitor

Brand:  Tocris (Bio-Techne) 042050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437633

  • £378.88 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent inhibitor of dopamine uptake (IC50 = 40 nM). Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

67469-45-8
521.474
gbr 13069 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylprop-2-en-1-yl piperazine dihydrochloride
1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride
C28H32Cl2F2N2O
ZBSZWEYIIGLGSX-RDRKJGRWSA-N
24978531
C1CN(CCN1CCOC(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F)CC=CC4=CC=CC=C4.Cl.Cl

Specifications

GBR 13069 dihydrochloride
67469-45-8
gbr 13069 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylprop-2-en-1-yl piperazine dihydrochloride
C1CN(CCN1CCOC(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F)CC=CC4=CC=CC=C4.Cl.Cl
521.474
521.48
50mg
C28H32Cl2F2N2O
ZBSZWEYIIGLGSX-RDRKJGRWSA-N
1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride
24978531
>98%
Videos
Documents