Learn More
GBR 12935 dihydrochloride, Tocris Bioscience™
Selective dopamine uptake inhibitor
Brand:  Tocris (Bio-Techne) 051450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487673
  • £231.01 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Potent and selective inhibitor of dopamine uptake (KD = 5.5 nM in rat striatal membranes).
Specifications
Chemical Identifiers
67469-81-2
487.509
1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
CHEBI:64091
C28H36Cl2N2O
NQWRSILGEXNJIT-UHFFFAOYSA-N
11957553
Specifications
GBR 12935 dihydrochloride
>98%
1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
487.509
50mg
487.51
67469-81-2
C28H36Cl2N2O
NQWRSILGEXNJIT-UHFFFAOYSA-N
1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
11957553
CHEBI:64091
Videos