Learn More
Tocris Bioscience™ GBR 12909 dihydrochloride
Selective DA uptake inhibitor; also σ ligand
Brand:  Tocris Bioscience™ 042110
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13447633
  • £95.55 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Potent, competitive inhibitor of dopamine uptake (Ki = 1 nM for inhibition of striatal dopamine uptake). Has > 100-fold lower affinity for the noradrenalin and 5-HT uptake carriers. Also a potent sigma ligand (IC50 = 48 nM). Centrally active following systemic administration.
Specifications
Specifications
1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride
>98%
Observed MW: 523.49
Soluble to 50mM in DMSO
67469-78-7
C28H32F2N2O.2HCl