missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GBR 12909 dihydrochloride
Selective DA uptake inhibitor; also σ ligand
POA
Specifications
Assay Percent Range | >98% |
---|---|
Chemical Name or Material | 1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride |
CAS | 67469-78-7 |
Description
Potent, competitive inhibitor of dopamine uptake (Ki = 1 nM for inhibition of striatal dopamine uptake). Has > 100-fold lower affinity for the noradrenalin and 5-HT uptake carriers. Also a potent sigma ligand (IC50 = 48 nM). Centrally active following systemic administration.Specifications
>98% | |
1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride | |
67469-78-7 |
For Research Use Only