Learn More
GBR 12783 dihydrochloride, Tocris Bioscience™
Potent and selective DA uptake inhibitor
POA
Description
Description
A very potent and selective inhibitor of dopamine uptake (IC50 for inhibition of [3H]-dopamine uptake in rat striatal synaptosomes is 1.8 nM).