missing translation for 'onlineSavingsMsg'
Learn More

GBR 12783 dihydrochloride, Tocris Bioscience™

Potent and selective DA uptake inhibitor

Brand:  Tocris (Bio-Techne) 051350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467673

  • £498.33 / 50mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A very potent and selective inhibitor of dopamine uptake (IC50 for inhibition of [3H]-dopamine uptake in rat striatal synaptosomes is 1.8 nM).
Specifications

Chemical Identifiers

67469-75-4
485.493
1-(2-Diphenylmethoxyethyl)-4-(3-phenyl-2-propenyl)-piperazine dihydrochloride
1-(2-benzhydryloxyethyl)-4-(3-phenylprop-2-enyl)piperazine;dihydrochloride
C28H34Cl2N2O
HJDXBLLTRMCYNC-UHFFFAOYSA-N
53422048
C1CN(CCN1CCOC(C2=CC=CC=C2)C3=CC=CC=C3)CC=CC4=CC=CC=C4.Cl.Cl

Specifications

50mg
67469-75-4
1-(2-Diphenylmethoxyethyl)-4-(3-phenyl-2-propenyl)-piperazine dihydrochloride
C1CN(CCN1CCOC(C2=CC=CC=C2)C3=CC=CC=C3)CC=CC4=CC=CC=C4.Cl.Cl
485.493
485.5
GBR 12783 dihydrochloride
C28H34Cl2N2O
HJDXBLLTRMCYNC-UHFFFAOYSA-N
1-(2-benzhydryloxyethyl)-4-(3-phenylprop-2-enyl)piperazine;dihydrochloride
53422048
>99%
Videos
Documents