missing translation for 'onlineSavingsMsg'
Learn More

Gavestinel, Tocris Bioscience™

Potent and selective glycine site antagonist; orally available and active in vivo

Brand:  Tocris (Bio-Techne) 234850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431084

  • £668.85 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent and selective non-competitive antagonist acting at the strychnine-insensitive glycine binding site of the NMDA receptor-channel complex (Kd = 0.8 nM). Displays > 1000-fold selectivity over NMDA, AMPA and kainate binding sites. Orally bioavailable and active in vivo.
Specifications

Chemical Identifiers

153436-38-5
398.195
gavestinel
3-[(E)-3-anilino-3-oxoprop-1-enyl]-4,6-dichloro-1H-indole-2-carboxylic acid;sodium
C18H12Cl2N2NaO3
OBPMZHMRJJJUBC-UHDJGPCESA-N
67241466
C1=CC=C(C=C1)NC(=O)C=CC2=C(NC3=CC(=CC(=C32)Cl)Cl)C(=O)O.[Na]

Specifications

50mg
153436-38-5
gavestinel
C1=CC=C(C=C1)NC(=O)C=CC2=C(NC3=CC(=CC(=C32)Cl)Cl)C(=O)O.[Na]
398.195
397.19
Gavestinel
C18H12Cl2N2NaO3
OBPMZHMRJJJUBC-UHDJGPCESA-N
3-[(E)-3-anilino-3-oxoprop-1-enyl]-4,6-dichloro-1H-indole-2-carboxylic acid;sodium
67241466
>98%
Videos
Documents