Learn More
Fritsch™ Gassing Lids
Brand:  Fritsch™ 50.8700.00
Code : NEW
Additional Details : Weight : 1.30000kg
Product Code. 10740493
  • POA / Each
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
Specifications
Specifications
80mL Volume Grinding Bowl
Tempered Steel
Gassing Lid
2 valves and seal ring
For Planetary Mono Mill Pulverisette 6 Classic Line
Videos