Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottle Head without Filter Disc
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottle Heads without Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05500600
Code : A0
Additional Details : Weight : 0.05400kg
Description
- Acc. to DIN 12596, with cone
- For bottles acc. to DIN 12463
- Corrosion-free and resistant to thermal shock
Specifications
Duran Tubing | |
Glass Olives | |
11 mm | |
1 Pack |
Gas Washing Bottle Head without Filter Disc | |
NS 29/32 | |
DIN 12596 |