Learn More
Cowie Technology™ Octahedral PTFE Stir Bar
Manufactured from PTFE encapsulated Alnico V magnet core. Fisherbrand™ Octahedral PTFE Stir Bar is autoclavable.
POA
Specifications
Autoclavable | Yes |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Color | White |
Description | PTFE encapsulated Alnico V magnet core |
Diameter (English) | 5/16 in. |
Description
Manufactured from PTFE encapsulated Alnico V magnet core. Fisherbrand™ Octahedral PTFE Stir Bar is autoclavable.
- Stable, general-purpose magnetic stir bar
- The flat surfaces provide a greater mixing action
- All have pivot rings
- AlNiCo-V magnets encased in PTFE
- Autoclavable
Specifications
Yes | |
White | |
5/16 in. | |
5/Pk. | |
Containers & Vessels / Stirrer Hot Plates |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
8 mm | |
Octahedral |