Learn More
Integra Biosciences™ Gas Cartridge Adapter
Equipped with this gas cartridge adapter, the FIREBOY is independent of any gas distribution system and power supply. Integra Biosciences™ Gas Cartridge Adapters allow FIREBOY burners to be used wherever required.
Brand:  Integra Biosciences™ 144050
Code : 04
Additional Details : Weight : 1.00000kg
Product Code. 11682080
  • £149.00 / Each
In Stock 3
Add to basket
Description
Description
  • Campingaz CV360 gas cartridge adapter with quick coupling
  • For FIREBOY models 144000 and 144010
Specifications
Specifications
52 G Butane Cartridge
Campingaz CV 360, Fireboy Bunsen Burner
1 Each
Adapter