missing translation for 'onlineSavingsMsg'
Learn More

GARP, APC, clone: YGIC86, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9891-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15310850

  • £80.93 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predominant (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta.When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells.YGIC86 does not cross-react to rat GARP.Applications Reported: This YGIC86 antibody has been reported for use in flow cytometric analysis.Applications Tested: This YGIC86 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

GARP
YGIC86
APC
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Lrrc32, Garpin
IgG2a
RUO
Primary
Murine
Videos