Learn More
GARP Monoclonal Antibody (G14D9), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£304.00
Specifications
Antigen | GARP |
---|---|
Clone | G14D9 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15519466
|
Affymetrix eBioscience
46-9882-42 |
100 Tests |
£304.00
100 tests |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Description: The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predomit (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta. Applications Reported: This G14D9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G14D9 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
GARP Monoclonal antibody specifically detects GARP in Human samples. It is validated for Flow Cytometry.Especificaciones
GARP | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q14392 | |
LRRC32 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
LRRC32 |
G14D9 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Human | |
2615 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.