Learn More
GARP Monoclonal Antibody (G14D9), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 25-9882-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predomit (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta. Applications Reported: This G14D9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G14D9 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
GARP Monoclonal antibody specifically detects GARP in Human samples. It is validated for Flow Cytometry.Specifications
GARP | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q14392 | |
LRRC32 | |
A mixture of mGARP-Fc and hGARP 21H-F1 & -F2 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
G14D9 | |
PE-Cyanine7 | |
LRRC32 | |
AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32 | |
Rat | |
Affinity chromatography | |
RUO | |
2615 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only.