missing translation for 'onlineSavingsMsg'
Learn More

GARP, FITC, clone: G14D9, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9882-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596636

  • £81.17 / 25 tests


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predominant (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta.Applications Reported: This G14D9 antibody has been reported for use in flow cytometric analysis.Applications Tested: This G14D9 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

GARP
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q14392
D11S833E, GARP, garpin, glycoprotein A repetitions predominant, leucine-rich repeat-containing protein 32
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
G14D9
FITC
LRRC32
Conjugated
LRRC32
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2615
Videos

For Research Use Only.