missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ GAP43 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human GAP43 (Met 1-Ala 238) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 12408H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15295905

  • £389.95 / 25µg


Estimated Shipment
04-07-2018
Add to basket

Description

Description

  • Formulation: 10mM Na2HPO4, 1.8mM KH2PO4, 137mM NaCl, 2.7mM KCl, 0.1mM EDTA, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2596
≥90% by SDS-PAGE
Human
<1 EU/μg
GAP43
Other Proteins
GAP43 Recombinant Human Protein
Lyophilized
26.2kd
5 x 5μg
Full Length, Recombinant
Growth Associated Protein 43
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.