missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ GAP43 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human GAP43 (Met 1-Ala 238) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 12408H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15225915

  • £649.59 / 50µg


Estimated Shipment
17-05-2018
Add to basket

Description

Description

  • Formulation: 10mM Na2HPO4, 1.8mM KH2PO4, 137mM NaCl, 2.7mM KCl, 0.1mM EDTA, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2596
≥90% by SDS-PAGE
Human
<1 EU/μg
GAP43
Other Proteins
GAP43 Recombinant Human Protein
Lyophilized
26.2kd
50μg
Full Length, Recombinant
Growth Associated Protein 43
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.