missing translation for 'onlineSavingsMsg'
Learn More

GANT 58, Tocris Bioscience™

GLI1 antagonist; inhibits Hedgehog (Hh) signaling

Brand:  Tocris (Bio-Techne) 388910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463764

  • £151.80 / 10mg


Estimated Shipment
02-01-2019
Add to basket

Description

Description

GLI antagonist that inhibits GLI1-induced transcription (IC50 = 5μM). Inhibits the hedgehog (Hh) signaling pathway downstream of SMO and SUFU causing GLI1 nuclear accumulation. Displays antiproliferative and antitumor activity in vivo.
Specifications

Chemical Identifiers

64048-12-0
392.48
gant 58, gant58, 2,3,4,5-tetra 4-pyridyl thiophene, 4-2,4,5-tri 4-pyridinyl-3-thienyl pyridine, 4-2,4,5-tripyridin-4-ylthiophen-3-yl pyridine, 4,4',4,4'-thiophene-2,3,4,5-tetrayl tetrapyridine, ncistruc1_001570, ncistruc2_001490, 4,4 ,4 ,4-2,3,4,5-thiophentetrayl tetrakis-pyridine, gant hplc
4-(2,4,5-tripyridin-4-ylthiophen-3-yl)pyridine
C24H16N4S
USWLOKMMUTWFMD-UHFFFAOYSA-N
253078
C1=CN=CC=C1C2=C(SC(=C2C3=CC=NC=C3)C4=CC=NC=C4)C5=CC=NC=C5

Specifications

GANT 58
64048-12-0
gant 58, gant58, 2,3,4,5-tetra 4-pyridyl thiophene, 4-2,4,5-tri 4-pyridinyl-3-thienyl pyridine, 4-2,4,5-tripyridin-4-ylthiophen-3-yl pyridine, 4,4',4,4'-thiophene-2,3,4,5-tetrayl tetrapyridine, ncistruc1_001570, ncistruc2_001490, 4,4 ,4 ,4-2,3,4,5-thiophentetrayl tetrakis-pyridine, gant hplc
C1=CN=CC=C1C2=C(SC(=C2C3=CC=NC=C3)C4=CC=NC=C4)C5=CC=NC=C5
392.48
392.48
10mg
C24H16N4S
USWLOKMMUTWFMD-UHFFFAOYSA-N
4-(2,4,5-tripyridin-4-ylthiophen-3-yl)pyridine
253078
>99%
Videos