Learn More
TCR gamma/delta Monoclonal Antibody (B1.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£101.00 - £250.00
Specifications
Antigen | TCR gamma/delta |
---|---|
Clone | B1.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15509476
|
Affymetrix eBioscience
46-9959-41 |
25 Tests |
£101.00
25 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15519476
|
Affymetrix eBioscience
46-9959-42 |
100 Tests |
£250.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The B1.1 monoclonal antibody reacts with the human gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection. This monoclonal can be used as a phenotypic marker for gamma delta TCR-expressing T cells. Applications Reported: This B1.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B1.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (2.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.Specifications
TCR gamma/delta | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
TRD, Trg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
B1.1 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
0 | |
6964, 6965 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.