missing translation for 'onlineSavingsMsg'
Learn More

TCR gamma/delta, clone: B1.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9959-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268277

  • £31.01 / 25µg


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Description: The B1.1 monoclonal antibody reacts with the human gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection. This monoclonal can be used as a phenotypic marker for gamma delta TCR-expressing T cells.Applications Reported: The B1.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The B1.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

gamma delta TCR
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Purified
TRD
IgG1, kappa
25μg
Store at 2-8°C.
Primary
6964
Flow Cytometry
B1.1
Unconjugated
TRD
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos

For Research Use Only.