missing translation for 'onlineSavingsMsg'
Learn More

TCR gamma/delta, FITC, clone: UC7-13D5, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5811-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15208269

  • £49.99 / 50µg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: The UC7-13D5 monoclonal antibody reacts with the mouse gammadelta TCR complex. gammadelta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gammadelta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection.Applications Reported: UC7-13D5 has been reported for use as a phenotypic marker for gamma delta TCR-expressing T cells in flow cytometric analysis.Applications Tested: The UC7-13D5 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

gamma delta TCR
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Conjugated
Tcrd
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
110066
Flow Cytometry
UC7-13D5
FITC
Tcrd
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.