missing translation for 'onlineSavingsMsg'
Learn More

TCR gamma/delta, FITC, clone: eBioGL3 (GL-3, GL3), eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5711-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15288239

  • £55.31 / 50µg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The GL3 monoclonal antibody (mAb) reacts with the gamma delta T-cell Receptor complex (TCR) on all gamma delta TCR bearing T cells, but it does not react with alpha beta TCR. The gamma delta TCR is present on cells in the thymus, epidermis, epithelial lining of the intestine, peritoneal cavity, and lymphoid tissues.Applications Reported: This GL3 (GL-3) antibody has been reported for use in flow cytometric analysis.It is recommended to prestain cells with anti-mouse CD16/32 (cat. 14-0161) to prevent non-specific Fc-mediated binding of the GL3 monoclonal antibody.Applications Tested: This eBioGL3 (GL-3, GL3) antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

gamma delta TCR
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Conjugated
Tcrd
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
110066
Flow Cytometry
eBioGL3 (GL-3, GL3)
FITC
Tcrd
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.