Learn More
Tocris Bioscience™ Gambogic acid
Apoptosis inducer. Activates caspases and inhibits Bcl-2 family proteins
Brand:  Tocris Bioscience™ 359050
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463254
  • £336.61 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Apoptosis inducer in several tumor cell lines including T47D cells. Activates caspases (EC50 values are 0.78 - 1.64μM) and competitively inhibits antiapoptotic Bcl-2 family proteins (IC50 values are 1.47, 1.21, 2.02, 0.66, 1.06 and 0.79μM for Bcl-XL, Bcl-2, Bcl-W, Bcl-B, Bfl-1 and Mcl-1 respectively). Reverses gefitinib-resistance in mice bearing tumors with EGFR-T790M mutation. Also blocks Kir2.1 channels (EC50 ≤ 100 nM).
Specifications
Specifications
Gambogic acid
Gambogic acid
628.75