missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00008693-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MAVRWTWAGKSCLLLAFLTVAYIFVELLVSTFHASAGAGRARELGSRRLSGLQKNTEDLSRPLYKKPPADSRALGEWGKASKLQLNEDELKQQEELIERYAINIYLSDRISLHRHIEDKRMYECKSQKFNYRTLPTTSVIIAFYNEAWSTLLRTIHSVLETSPAVLLKEIILVDDLSDRVYLKTQLETYISNLDRVRLIRTNKREGLVRARLIGATFATGDVLTFLDCHCECNSGWLEPLLERIGRDETAVVCPVIDTIDWNTFEFYMQIGEPMIGGFDWRLTFQWHSVPKQERDRRISRIDPIRSPTMAGGLFAVSKKYFQYLGTYDTGMEVWGGENLELSFRVWQCGGKLEIHPCSHVGHVFPKRAPYARPNFLQNTARAAEVWMDEYKEHFYNRNPPARKEAYGDISERKLLRERLRCKSFDWYLKNVFPNLHVPEDRPGWHGAIRSRGISSECLDYNSPDNNPTGANLSLFGCHGQGGNQFFEYTSNKEIRFNSVTELCAEVPEQKNYVGMQNCPKDGFPVPANIIWHFKEDGTIFHPHSGLCLSAYRTPEGRPDVQMRTCDALDKNQIWSFEKSpecifications
GALNT4 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC036390.2 | |
GALNT4 | |
GALNT4 (AAH36390.1, 1 a.a. ∼ 578 a.a) full-length human protein. | |
RUO | |
8693 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4 (GalNAc-T4) | |
GALNAC-T4/GalNAcT4 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody |