missing translation for 'onlineSavingsMsg'
Learn More

Gallocyanine, Thermo Scientific Chemicals

Catalog Number p-2369402
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1562-85-2
C15H13ClN2O5
336.72
MFCD00011926
AQSOTOUQTVJNMY-UHFFFAOYSA-N
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
73801
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]
This item is not returnable. View return policy
1562-85-2
C15H13ClN2O5
336.72
MFCD00011926
AQSOTOUQTVJNMY-UHFFFAOYSA-N
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
73801
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Green
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 27, 438
Merck Index 15, 4385
Formula Weight 336.72
Physical Form Fine Crystalline Powder
Chemical Name or Material Gallocyanine
compliance-icons
Product Identifier
  • Gallocyanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'