missing translation for 'onlineSavingsMsg'
Learn More

Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11303427
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
7440-31-5, 7440-55-3
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
This item is not returnable. View return policy
7440-31-5, 7440-55-3
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]

Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Melting Point ≥25°C
Appearance Silver-colored liquid
Assay Percent Range 99.99% (metals basis)
Quantity 1 g
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Tin: 8.0%; Gallium: 92.0%
Form Melting Solid
Chemical Name or Material Gallium Tin alloy

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'