Learn More
Gallium Indium alloy, 99.99% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 044240.09
92.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Step soldering involving temperature sensitive components, Soldering to molded interconnect device (MID) plastics, Fusible alloys/fuse applications, Mercury replacement, Thermal and electrical conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
99.99% (metals basis) | |
MFCD00144387 | |
Insoluble in water. | |
Solid |
Gallium Indium alloy | |
Ga:In; 40:60 wt.% | |
10 g | |
Odorless |
RUO – Research Use Only