Learn More
Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals
Description
Gallium(III) nitrate hydrate is used in the preparation of gallium nitride and other gallium compounds. It acts as a drug used to treat symptomatic hypercalcemia secondary to cancer. It finds application as an antineoplastic agent and also as an acting agent for the treatment of bone-resorptive diseases. It is involved to improve the cultural selectivity of medically significant fungi by suppression of contaminating bacterial flora.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 110°C (decomposition) |
Color | White |
Physical Form | Lump |
Quantity | 5 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | Soluble in water. |
IUPAC Name | gallium(3+) trinitrate |
Formula Weight | 255.73 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only