missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) isopropoxide, mixture of oligomers, 99%, Thermo Scientific Chemicals

Catalog Number p-7050356
Click to view available options
Quantity:
1 g
5 g
4452-61-3
C3H8GaO
129.819
MFCD00144389
JTPVILBGRKEEOP-UHFFFAOYSA-N
2-propanol, gallium salt
122197675
gallium;propan-2-ol
CC(C)O.[Ga]
4452-61-3
C3H8GaO
129.819
MFCD00144389
JTPVILBGRKEEOP-UHFFFAOYSA-N
2-propanol, gallium salt
122197675
gallium;propan-2-ol
CC(C)O.[Ga]

Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 72°C
Color Yellow
Density 1.35 g/mL
Boiling Point 120°C (1 mm)
Flash Point 68°C (154°F)
Linear Formula Ga(OC3H7)3
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 246.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Gallium(III) isopropoxide, Mixture of oligomers
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'