missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) fluoride, anhydrous, 99.85% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049965
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7783-51-9
MFCD00011021
This item is not returnable. View return policy
7783-51-9
MFCD00011021

Gallium(III) fluoride reacts with mineral acids to give hydrofluoric acid. It is used in the preparation and study of optical fluorogallate glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼1000°C
Melting Point 800°C (sublimation)
Color White
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4354
Formula Weight 126.72
Percent Purity 99.85%
Odor Odorless
Sensitivity Moisture Sensitive
Density 4.47g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'