missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gallium(III) ethoxide

Brand:  Alfa Aesar™ 044859.14

Code : NEW

Additional Details : CAS Number : 2572-25-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11316279

  • £881.80 / 25g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Gallium (III) ethoxide
C6H15GaO3·xHOCH2CH3
204.9
Gallium triethoxide
2572-25-0
Ga(OC2H5)3·xHOCH2CH3
MFCD01866730

Specifications

Gallium (III) ethoxide
25g
Liquid
C6H15GaO3·xHOCH2CH3
204.9
Gallium triethoxide
68°C (154°F)
Moisture Sensitive
2572-25-0
1.25g/mL
Ga(OC2H5)3·xHOCH2CH3
MFCD01866730
Yellow
Videos
SDS