missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium(III) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Gallium bromide is used as a catalyst in the alkylation of aromatic compounds by alkyl bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 279°C |
Melting Point | 122°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
UN Number | UN3260 |
Solubility Information | Soluble in water. |
Formula Weight | 309.45 |
Percent Purity | 99.998% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'