missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 253.0°C |
Color | Yellow to Beige |
Assay Percent Range | 98% |
Solubility Information | Solubility in water: 12g/L cold water. Other solubilities: easily soluble in warmed water, very soluble in ethanol: 160g/L, acetone: 200g/l, soluble in glycerol: 100g/L, slightly soluble in, ether: 10g/L, insoluble in benzene, chloroform, petrol ether |
Formula Weight | 170.12 |
Percent Purity | 98% |
Density | 1.6940g/mL |
Quantity | 1 kg |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'