missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Galectin-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Galectin-4 Polyclonal specifically detects Galectin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Galectin-4 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, Galectin4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4, LGALS4 | |
LGALS4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q8K419 | |
3960 | |
E. coli-derived recombinant mouse Galectin-4 Ala2-Ile326 Accession # Q8K419 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Galectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human (rh) Galectin-4, approximately 5% cross-reactivity with recombinant mouse (rm) Galectin-3, rmGalectin-7, and rhGalectin-8, and less than 1% cross-reactivity with rmGalectin-1 and rhGalectin-2 is observed. |
For Research Use Only