Learn More
Galectin 3 Monoclonal Antibody (eBioM3/38 (M3/38)), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£66.50 - £219.00
Specifications
Antigen | Galectin 3 |
---|---|
Clone | eBioM3/38 (M3/38) |
Concentration | 0.5 mg/mL |
Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15287407
|
Affymetrix eBioscience
14-5301-81 |
50 μg |
£66.50
50µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15297407
|
Affymetrix eBioscience
14-5301-82 |
100 μg |
£133.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15207417
|
Affymetrix eBioscience
14-5301-85 |
500 μg |
£219.00
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The eBioM3/38 antibody reacts with mouse Galectin-3 (Mac-2). Galectins are a family of animal lectins which appear to exhibit a variety of biological functions. Galectin 3 (aka LGALS3, galactose-specific soluble lectin 3, Mac-2, L-29) is one of the more extensively studied members of this family and is a 30 kDa beta-galactoside-binding protein. Due to a C-terminal carbohydrate binding site, Galectin 3 is capable of binding IgE and mammalian cell surfaces only when homodimerized or homo-oligomerized. Galectin 3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. This monoclonal antibody eBioM3/38 crossreacts to human Galectin-3 (LGALS3). Applications Reported: This eBioM3/38 (M3/38) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), immunohistology staining of frozen tissue sections, immunohistology staining of paraffin embedded tissue sections, and ELISA. Applications Tested: This eBioM3/38 (M3/38) antibody has been tested by flow cytometric analysis of mouse PECs (thioglycollated elicied peritioneal cells). This can be used at less than or equal to 0.25 μg per test.
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.Specifications
Galectin 3 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
35 kDa lectin; advanced glycation end-product receptor 3; AGE-R3; beta-galactosides-binding lectin; carbohydrate-binding protein 35; CBP 35; CBP30; CBP-30; CBP35; epsilon BP; gal3; gal-3; galactose-specific lectin 3; Galactoside-binding protein; GALBP; galectin 3; Galectin3; galectin-3; GALIG; GBP; IgE binding protein; igE-binding protein; L-29; L31; L-31; L-34; L-34 galactoside-binding lectin; Laminin-binding protein; lectin galactoside-binding soluble 3; lectin galactoside-binding soluble 3 protein; lectin L-29; lectin, galactose binding, soluble 3; lectin, galactoside binding soluble 3; lectin, galactoside-binding, soluble, 3; lectin, galactoside-binding, soluble, 3 (galectin 3); LGALS2; Lgals3; MAC2; Mac-2; Mac-2 antigen | |
LGALS3 | |
Primary | |
4° C | |
LGALS3 |
eBioM3/38 (M3/38) | |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
P16110, P17931 | |
16854, 3958 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.